This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05144333_g1
See other LOC101698172 GE Assays ›
Gene Symbol:
LOC101698172
Gene Name:
histone H3.3
Gene Aliases:
GW7_02941, H3f3b
Chromosome Location:
Chr.Un|NW_004624801: 5668661 - 5672073 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05144333_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101698172
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004860706.3 XP_004860763.1 1 - 2 194 61
XM_013078181.2 XP_012933635.1 1 - 2 194 61
XM_013078182.2 XP_012933636.1 1 - 2 194 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004860706.3, XM_013078181.2, XM_013078182.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

histone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline