This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05147502_m1
See other ATP23 GE Assays ›
Gene Symbol:
Atp23
Gene Name:
ATP23 metallopeptidase and ATP synthase assembly factor homolog
Gene Aliases:
GW7_04663
Chromosome Location:
Chr.Un|NW_004624802: 9994792 - 10010822 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05147502_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004861157.2 XP_004861214.1 4 - 5 642 96
XM_004861158.2 XP_004861215.1 4 - 5 642 96
XM_021241468.1 XP_021097127.1 4 - 5 664 96
XM_021241469.1 XP_021097128.1 2 - 3 209 96
XM_021241470.1 XP_021097129.1 3 - 4 268 96
XM_021241471.1 XP_021097130.1 3 - 4 293 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline