This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05159909_g1
See other TBRG4 GE Assays ›
Gene Symbol:
Tbrg4
Gene Name:
transforming growth factor beta regulator 4
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624740: 7277260 - 7287073 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05159909_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tbrg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004839888.2 XP_004839945.1 4 - 5 1091 52
XM_004839889.2 XP_004839946.1 4 - 5 1110 52
XM_004839890.2 XP_004839947.1 4 - 5 983 52
XM_004839891.3 XP_004839948.1 4 - 5 1457 52
XM_021254260.1 XP_021109919.1 4 - 5 1087 52
XM_021254261.1 XP_021109920.1 4 - 5 983 52
XM_021254263.1 XP_021109922.1 4 - 5 1040 52

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline