This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05174021_g1
See other ESRRG GE Assays ›
Gene Symbol:
Esrrg
Gene Name:
estrogen related receptor gamma
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624835: 816516 - 1015687 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05174021_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Esrrg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867386.3 XP_004867443.1 5 - 6 986 66
XM_013066530.2 XP_012921984.1 6 - 7 1270 66
XM_013066531.2 XP_012921985.1 6 - 7 1047 66
XM_021246443.1 XP_021102102.1 5 - 6 1013 66
XM_021246444.1 XP_021102103.1 5 - 6 960 66
XM_021246445.1 XP_021102104.1 5 - 6 876 66
XM_021246446.1 XP_021102105.1 5 - 6 1105 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0