This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05174135_m1
See other DSE GE Assays ›
Gene Symbol:
Dse
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
GW7_20796
Chromosome Location:
Chr.Un|NW_004624856: 2016465 - 2100096 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05174135_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dse
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004869072.2 XP_004869129.1 4 - 5 1066 78
XM_004869073.2 XP_004869130.1 4 - 5 1088 78
XM_013067192.2 XP_012922646.1 3 - 4 680 78
XM_013067193.2 XP_012922647.1 3 - 4 661 78
XM_021247825.1 XP_021103484.1 3 - 4 809 78
XM_021247826.1 XP_021103485.1 4 - 5 1067 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9mkk8:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0