This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05175561_g1
See other MUTYH GE Assays ›
Gene Symbol:
Mutyh
Gene Name:
mutY DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624906: 1238332 - 1247813 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05175561_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mutyh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021250809.1 XP_021106468.1 3 - 4 372 60
XM_021250810.1 XP_021106469.1 3 - 4 372 60
XM_021250811.1 XP_021106470.1 3 - 4 419 60
XM_021250812.1 XP_021106471.1 3 - 4 413 60
XM_021250814.1 XP_021106473.1 3 - 4 729 60
XM_021250815.1 XP_021106474.1 2 - 3 205 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021250815.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0