This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05177414_mH
See other OPCML GE Assays ›
Gene Symbol:
Opcml
Gene Name:
opioid binding protein/cell adhesion molecule like
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624812: 6808915 - 8042941 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05177414_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Opcml
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004862648.2 XP_004862705.1 5 - 6 1429 69
XM_004862649.3 XP_004862706.1 6 - 7 869 69
XM_004862651.2 XP_004862708.1 5 - 6 1428 69
XM_004862652.2 XP_004862709.1 5 - 6 1426 69
XM_021242762.1 XP_021098421.1 5 - 6 1431 69
XM_021242763.1 XP_021098422.1 5 - 6 1430 69
XM_021242764.1 XP_021098423.1 4 - 5 1307 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline