This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05179512_g1
See other P2RX7 GE Assays ›
Gene Symbol:
P2rx7
Gene Name:
purinergic receptor P2X 7
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624747: 21725808 - 21768049 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05179512_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004843831.3 XP_004843888.1 2 - 3 438 70
XM_004843833.3 XP_004843890.1 2 - 3 437 70
XM_021257501.1 XP_021113160.1 2 - 3 289 70
XM_021257502.1 XP_021113161.1 2 - 3 440 70
XM_021257503.1 XP_021113162.1 2 - 3 439 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0