This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05180425_m1
See other NUDT5 GE Assays ›
Gene Symbol:
Nudt5
Gene Name:
nudix hydrolase 5
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624805: 4903250 - 4922699 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05180425_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004861813.2 XP_004861870.1 6 - 7 605 67
XM_013078468.2 XP_012933922.1 7 - 8 701 67
XM_013078470.2 XP_012933924.1 7 - 8 760 67
XM_021242068.1 XP_021097727.1 8 - 9 846 67
XM_021242070.1 XP_021097729.1 9 - 10 1103 67
XM_021242071.1 XP_021097730.1 8 - 9 906 67
XM_021242072.1 XP_021097731.1 8 - 9 957 67
XM_021242073.1 XP_021097732.1 7 - 8 748 67
XM_021242074.1 XP_021097733.1 6 - 7 662 67
XM_021242075.1 XP_021097734.1 7 - 8 703 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

pyrophosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0