This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05182613_m1
See other ARFGAP1 GE Assays ›
Gene Symbol:
Arfgap1
Gene Name:
ADP ribosylation factor GTPase activating protein 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624741: 28971334 - 28986717 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05182613_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arfgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004841043.3 XP_004841100.1 7 - 8 778 65
XM_004841044.3 XP_004841101.1 7 - 8 778 65
XM_004841045.3 XP_004841102.1 7 - 8 778 65
XM_004841046.3 XP_004841103.1 7 - 8 778 65
XM_004841047.3 XP_004841104.1 7 - 8 778 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0