This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05186170_g1
See other PGGHG GE Assays ›
Gene Symbol:
Pgghg
Gene Name:
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624766: 22136826 - 22142601 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05186170_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgghg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004851750.3 XP_004851807.1 5 - 6 1399 66
XM_004851756.3 XP_004851813.1 5 - 6 1399 66
XM_004851757.3 XP_004851814.1 5 - 6 1399 66
XM_004851758.3 XP_004851815.1 5 - 6 1399 66
XM_013074669.2 XP_012930123.1 5 - 6 1075 66
XM_021264088.1 XP_021119747.1 4 - 5 1381 66
XM_021264089.1 XP_021119748.1 5 - 6 1399 66
XM_021264090.1 XP_021119749.1 5 - 6 1399 66
XM_021264091.1 XP_021119750.1 5 - 6 1399 66
XM_021264092.1 XP_021119751.1 5 - 6 1057 66
XM_021264093.1 XP_021119752.1 5 - 6 1399 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0