This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05186258_m1
See other CABLES1 GE Assays ›
Gene Symbol:
Cables1
Gene Name:
Cdk5 and Abl enzyme substrate 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624770: 8837217 - 8935789 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05186258_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cables1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004853161.3 XP_004853218.1 2 - 3 791 80
XM_004853162.3 XP_004853219.1 2 - 3 791 80
XM_021265158.1 XP_021120817.1 2 - 3 795 80
XM_021265159.1 XP_021120818.1 2 - 3 800 80
XM_021265160.1 XP_021120819.1 2 - 3 197 80
XM_021265161.1 XP_021120820.1 2 - 3 158 80
XM_021265162.1 XP_021120821.1 2 - 3 94 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021265161.1, XM_021265162.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0