This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05192327_g1
See other GCNT3 GE Assays ›
Gene Symbol:
Gcnt3
Gene Name:
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624781: 12522318 - 12525089 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05192327_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcnt3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855724.3 XP_004855781.2 2 - 3 302 70
XM_021237136.1 XP_021092795.1 2 - 3 145 70
XM_021237137.1 XP_021092796.1 2 - 3 368 70
XM_021237138.1 XP_021092797.1 3 - 4 397 70
XM_021237139.1 XP_021092798.1 2 - 3 1467 70
XM_021237140.1 XP_021092799.1 2 - 3 404 70
XM_021237141.1 XP_021092800.1 2 - 3 211 70
XM_021237143.1 XP_021092802.1 3 - 4 331 70
XM_021237144.1 XP_021092803.1 2 - 3 1401 70
XM_021237145.1 XP_021092804.1 2 - 3 338 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline