This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05192328_g1
See other GCNT3 GE Assays ›
Gene Symbol:
Gcnt3
Gene Name:
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624781: 12522318 - 12525089 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05192328_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcnt3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855724.3 XP_004855781.2 3 - 4 335 95
XM_021237136.1 XP_021092795.1 3 - 4 178 95
XM_021237137.1 XP_021092796.1 3 - 4 401 95
XM_021237138.1 XP_021092797.1 4 - 5 430 95
XM_021237139.1 XP_021092798.1 3 - 4 1500 95
XM_021237140.1 XP_021092799.1 3 - 4 437 95
XM_021237141.1 XP_021092800.1 3 - 4 244 95
XM_021237143.1 XP_021092802.1 4 - 5 364 95
XM_021237144.1 XP_021092803.1 3 - 4 1434 95
XM_021237145.1 XP_021092804.1 3 - 4 371 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0