This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05196602_m1
See other LIPE GE Assays ›
Gene Symbol:
Lipe
Gene Name:
lipase E, hormone sensitive type
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624907: 363515 - 379470 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05196602_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004873010.3 XP_004873067.1 7 - 8 1552 65
XM_013068617.2 XP_012924071.1 7 - 8 2551 65
XM_013068618.2 XP_012924072.1 7 - 8 1687 65
XM_013068619.2 XP_012924073.1 7 - 8 1957 65
XM_013068620.2 XP_012924074.1 7 - 8 1640 65
XM_013068621.2 XP_012924075.2 7 - 8 2058 65
XM_021250883.1 XP_021106542.1 7 - 8 2416 65
XM_021250884.1 XP_021106543.1 7 - 8 5317 65
XM_021250885.1 XP_021106544.1 7 - 8 1822 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

lipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0