This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05198045_m1
See other TFDP2 GE Assays ›
Gene Symbol:
Tfdp2
Gene Name:
transcription factor Dp-2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624730: 18562721 - 18704317 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05198045_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfdp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004834367.3 XP_004834424.1 3 - 4 270 105
XM_004834368.3 XP_004834425.1 2 - 3 274 105
XM_004834370.3 XP_004834427.1 2 - 3 270 105
XM_004834371.3 XP_004834428.1 2 - 3 196 105
XM_013071028.2 XP_012926482.1 3 - 4 243 105
XM_013071047.2 XP_012926501.1 2 - 3 203 105
XM_013071048.2 XP_012926502.1 2 - 3 126 105
XM_021251972.1 XP_021107631.1 4 - 5 571 105
XM_021252086.1 XP_021107745.1 3 - 4 270 105
XM_021252126.1 XP_021107785.1 3 - 4 270 105
XM_021252143.1 XP_021107802.1 4 - 5 252 105
XM_021252184.1 XP_021107843.1 3 - 4 270 105
XM_021252218.1 XP_021107877.1 3 - 4 270 105
XM_021252372.1 XP_021108031.1 2 - 3 225 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004834370.3, XM_004834371.3, XM_013071047.2, XM_013071048.2, XM_021252372.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0