This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05203664_m1
See other GNGT2 GE Assays ›
Gene Symbol:
Gngt2
Gene Name:
G protein subunit gamma transducin 2
Gene Aliases:
GW7_21755
Chromosome Location:
Chr.Un|NW_004624795: 5444335 - 5448529 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05203664_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gngt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021240056.1 XP_021095715.1 2 - 3 198 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021240056.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline