This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05206968_g1
See other KNL1 GE Assays ›
Gene Symbol:
Knl1
Gene Name:
kinetochore scaffold 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624804: 7893723 - 7952523 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05206968_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Knl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013078456.2 XP_012933910.1 13 - 14 5590 65
XM_021241892.1 XP_021097551.1 13 - 14 5656 65
XM_021241893.1 XP_021097552.1 13 - 14 5839 65
XM_021241894.1 XP_021097553.1 13 - 14 5804 65
XM_021241895.1 XP_021097554.1 13 - 14 5534 65
XM_021241896.1 XP_021097555.1 9 - 10 5371 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0