This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05211804_g1
See other RPUSD1 GE Assays ›
Gene Symbol:
Rpusd1
Gene Name:
RNA pseudouridylate synthase domain containing 1
Gene Aliases:
GW7_12965
Chromosome Location:
Chr.Un|NW_004624913: 1591311 - 1594467 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05211804_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpusd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004873836.3 XP_004873893.1 2 - 3 406 69
XM_021251344.1 XP_021107003.1 2 - 3 291 69
XM_021251345.1 XP_021107004.1 1 - 2 192 69
XM_021251346.1 XP_021107005.1 2 - 3 291 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021251345.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0