This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05219850_m1
See other ABL2 GE Assays ›
Gene Symbol:
Abl2
Gene Name:
ABL proto-oncogene 2, non-receptor tyrosine kinase
Gene Aliases:
GW7_01703
Chromosome Location:
Chr.Un|NW_004624771: 3160598 - 3278787 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05219850_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004853358.3 XP_004853415.1 11 - 12 2125 64
XM_004853359.2 XP_004853416.1 11 - 12 1827 64
XM_004853360.3 XP_004853417.1 10 - 11 2062 64
XM_004853361.3 XP_004853418.1 11 - 12 2124 64
XM_004853362.2 XP_004853419.1 11 - 12 1827 64
XM_004853363.3 XP_004853420.1 10 - 11 2061 64
XM_021265356.1 XP_021121015.1 11 - 12 2126 64
XM_021265357.1 XP_021121016.1 10 - 11 1853 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0