This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05220749_g1
See other PPP4R3B GE Assays ›
Gene Symbol:
Ppp4r3b
Gene Name:
protein phosphatase 4 regulatory subunit 3B
Gene Aliases:
Smek2
Chromosome Location:
Chr.Un|NW_004624833: 5935948 - 6009565 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05220749_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4r3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867259.2 XP_004867316.1 3 - 4 520 69
XM_013066445.2 XP_012921899.1 4 - 5 607 69
XM_013066446.2 XP_012921900.1 4 - 5 608 69
XM_021246298.1 XP_021101957.1 4 - 5 609 69
XM_021246300.1 XP_021101959.1 4 - 5 608 69
XM_021246301.1 XP_021101960.1 2 - 3 161 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0