Assay Details
| Gene Symbol: Pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843172.2 | XP_004843229.1 | 2 - 3 | 423 | 86 | |
| XM_021257018.1 | XP_021112677.1 | 2 - 3 | 427 | 86 | ||
More Information
Set Membership: |
|

Pllp
Genomic Map