This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05225880_m1
See other ATXN1 GE Assays ›
Gene Symbol:
Atxn1
Gene Name:
ataxin 1
Gene Aliases:
Ataxin-1, GW7_15391
Chromosome Location:
Chr.Un|NW_004624756: 9769331 - 10167560 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05225880_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atxn1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004847643.3 XP_004847700.1 8 - 9 2965 97
XM_004847648.3 XP_004847705.1 8 - 9 2932 97
XM_004847649.2 XP_004847706.1 7 - 8 2802 97
XM_013073219.2 XP_012928673.1 6 - 7 2654 97
XM_013073220.2 XP_012928674.1 8 - 9 2825 97
XM_013073221.2 XP_012928675.1 6 - 7 2711 97
XM_021260599.1 XP_021116258.1 7 - 8 2775 97
XM_021260600.1 XP_021116259.1 8 - 9 2954 97
XM_021260601.1 XP_021116260.1 8 - 9 2806 97
XM_021260602.1 XP_021116261.1 8 - 9 2905 97
XM_021260603.1 XP_021116262.1 8 - 9 2865 97
XM_021260604.1 XP_021116263.1 8 - 9 2621 97
XM_021260606.1 XP_021116265.1 8 - 9 2951 97
XM_021260607.1 XP_021116266.1 9 - 10 2856 97
XM_021260608.1 XP_021116267.1 8 - 9 2852 97
XM_021260609.1 XP_021116268.1 6 - 7 2660 97
XM_021260610.1 XP_021116269.1 6 - 7 2531 97
XM_021260611.1 XP_021116270.1 4 - 5 2460 97
XM_021260612.1 XP_021116271.1 3 - 4 2934 97
XM_021260614.1 XP_021116273.1 4 - 5 2551 97
XM_021260615.1 XP_021116274.1 3 - 4 2316 97
XM_021260616.1 XP_021116275.1 3 - 4 3075 97
XM_021260617.1 XP_021116276.1 4 - 5 2427 97
XM_021260618.1 XP_021116277.1 3 - 4 2407 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline