This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05226683_m1
See other HSD17B7 GE Assays ›
Gene Symbol:
Hsd17b7
Gene Name:
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624826: 1260223 - 1283866 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05226683_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd17b7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004865174.3 XP_004865231.1 5 - 6 733 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-5c659bcd8-pv2kk:80/100.66.76.15:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline