This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05227400_g1
See other RARG GE Assays ›
Gene Symbol:
Rarg
Gene Name:
retinoic acid receptor gamma
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624904: 1256931 - 1277947 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05227400_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rarg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013068487.2 XP_012923941.1 8 - 9 1590 74
XM_013068488.2 XP_012923942.2 7 - 8 2009 74
XM_021250595.1 XP_021106254.1 10 - 11 1962 74
XM_021250596.1 XP_021106255.1 10 - 11 1768 74
XM_021250597.1 XP_021106256.1 9 - 10 1851 74
XM_021250598.1 XP_021106257.1 9 - 10 1657 74
XM_021250599.1 XP_021106258.1 7 - 8 3290 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0