This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05230331_m1
See other NDUFB3 GE Assays ›
Gene Symbol:
Ndufb3
Gene Name:
NADH:ubiquinone oxidoreductase subunit B3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624889: 131731 - 141603 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05230331_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004871637.3 XP_004871694.1 3 - 4 514 89
XM_004871638.3 XP_004871695.1 3 - 4 509 89
XM_004871639.3 XP_004871696.1 2 - 3 418 89
XM_021249754.1 XP_021105413.1 3 - 4 522 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0