This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05232776_m1
See other AIMP1 GE Assays ›
Gene Symbol:
Aimp1
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Gene Aliases:
GW7_17850
Chromosome Location:
Chr.Un|NW_004624830: 2624310 - 2649099 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05232776_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aimp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004866233.2 XP_004866290.2 6 - 7 988 65
XM_004866234.2 XP_004866291.1 6 - 7 955 65
XM_004866235.3 XP_004866292.1 6 - 7 963 65
XM_021245454.1 XP_021101113.1 6 - 7 1811 65
XM_021245455.1 XP_021101114.1 6 - 7 820 65
XM_021245456.1 XP_021101115.1 5 - 6 846 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-976xq:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline