This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05243302_m1
See other PPIL3 GE Assays ›
Gene Symbol:
Ppil3
Gene Name:
peptidylprolyl isomerase like 3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624889: 283625 - 293755 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05243302_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004871645.2 XP_004871702.1 3 - 4 300 92
XM_004871647.2 XP_004871704.1 3 - 4 300 92
XM_021249763.1 XP_021105422.1 3 - 4 300 92
XM_021249764.1 XP_021105423.1 3 - 4 440 92
XM_021249765.1 XP_021105424.1 3 - 4 364 92
XM_021249766.1 XP_021105425.1 3 - 4 300 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline