This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05247674_g1
See other MRPL41 GE Assays ›
Gene Symbol:
Mrpl41
Gene Name:
mitochondrial ribosomal protein L41
Gene Aliases:
GW7_03933
Chromosome Location:
Chr.Un|NW_004624760: 517200 - 518784 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05247674_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021261593.1 XP_021117252.1 3 - 4 500 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021261593.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline