This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05251161_m1
See other PLAGL1 GE Assays ›
Gene Symbol:
Plagl1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624753: 10246498 - 10359073 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05251161_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plagl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004846028.2 XP_004846085.1 5 - 6 987 76
XM_004846029.3 XP_004846086.1 5 - 6 1064 76
XM_004846030.3 XP_004846087.1 5 - 6 911 76
XM_013072696.2 XP_012928150.1 6 - 7 1082 76
XM_013072697.2 XP_012928151.1 5 - 6 983 76
XM_013072698.2 XP_012928152.1 6 - 7 1078 76
XM_013072699.2 XP_012928153.1 5 - 6 762 76
XM_013072700.2 XP_012928154.1 5 - 6 775 76
XM_021259298.1 XP_021114957.1 5 - 6 686 76
XM_021259299.1 XP_021114958.1 5 - 6 839 76
XM_021259300.1 XP_021114959.1 5 - 6 699 76
XM_021259301.1 XP_021114960.1 6 - 7 857 76
XM_021259302.1 XP_021114961.1 6 - 7 853 76
XM_021259303.1 XP_021114962.1 5 - 6 758 76
XM_021259304.1 XP_021114963.1 6 - 7 870 76
XM_021259305.1 XP_021114964.1 5 - 6 771 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0