This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05257513_m1
See other PRSS55 GE Assays ›
Gene Symbol:
Prss55
Gene Name:
serine protease 55
Gene Aliases:
GW7_15146
Chromosome Location:
Chr.Un|NW_004624758: 24373667 - 24448596 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05257513_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004848441.2 XP_004848498.1 6 - 7 1650 75
XM_021261350.1 XP_021117009.1 3 - 4 844 75
XM_021261351.1 XP_021117010.1 6 - 7 1639 75
XM_021261352.1 XP_021117011.1 4 - 5 2428 75
XM_021261353.1 XP_021117012.1 6 - 7 1549 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline