This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05258113_g1
See other ARHGAP36 GE Assays ›
Gene Symbol:
Arhgap36
Gene Name:
Rho GTPase activating protein 36
Gene Aliases:
GW7_12767
Chromosome Location:
Chr.Un|NW_004624797: 10501784 - 10556386 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05258113_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004860066.2 XP_004860123.1 6 - 7 976 63
XM_004860068.2 XP_004860125.1 6 - 7 975 63
XM_004860069.2 XP_004860126.1 6 - 7 979 63
XM_004860070.2 XP_004860127.1 6 - 7 981 63
XM_004860071.3 XP_004860128.1 5 - 6 767 63
XM_021240609.1 XP_021096268.1 7 - 8 983 63
XM_021240610.1 XP_021096269.1 6 - 7 819 63
XM_021240611.1 XP_021096270.1 6 - 7 785 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline