This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05258734_m1
See other TRMT10B GE Assays ›
Gene Symbol:
Trmt10b
Gene Name:
tRNA methyltransferase 10B
Gene Aliases:
GW7_18486
Chromosome Location:
Chr.Un|NW_004624930: 1367368 - 1393009 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05258734_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trmt10b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004874586.2 XP_004874643.1 8 - 9 963 63
XM_004874589.3 XP_004874646.1 9 - 10 1088 63
XM_004874590.3 XP_004874647.1 8 - 9 905 63
XM_021251997.1 XP_021107656.1 8 - 9 958 63
XM_021251998.1 XP_021107657.1 9 - 10 1084 63
XM_021251999.1 XP_021107658.1 8 - 9 910 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline