This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05259239_m1
See other MAJIN GE Assays ›
Gene Symbol:
Majin
Gene Name:
membrane anchored junction protein
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624767: 21423175 - 21468227 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05259239_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Majin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013075043.2 XP_012930497.1 2 - 3 387 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-695c48667f-rs82x:80/100.66.76.254:80.
git-commit: c086736f0d622c41bbf3ea4d71b1c2b8619a5b9d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.35.0-Offline