This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05260235_m1
See other MIER1 GE Assays ›
Gene Symbol:
Mier1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624742: 28045141 - 28100683 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05260235_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mier1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004841483.3 XP_004841540.1 14 - 15 1415 76
XM_004841484.3 XP_004841541.1 12 - 13 1303 76
XM_013070968.2 XP_012926422.1 12 - 13 1334 76
XM_013070969.2 XP_012926423.1 14 - 15 1432 76
XM_013070970.2 XP_012926424.1 11 - 12 1242 76
XM_013070971.2 XP_012926425.1 13 - 14 1395 76
XM_013070972.2 XP_012926426.2 11 - 12 1403 76
XM_013070973.2 XP_012926427.1 13 - 14 1365 76
XM_013070974.2 XP_012926428.1 11 - 12 1197 76
XM_021255569.1 XP_021111228.1 14 - 15 1446 76
XM_021255570.1 XP_021111229.1 14 - 15 1407 76
XM_021255571.1 XP_021111230.1 13 - 14 1392 76
XM_021255572.1 XP_021111231.1 12 - 13 1312 76
XM_021255573.1 XP_021111232.1 12 - 13 1308 76
XM_021255574.1 XP_021111233.1 11 - 12 1109 76
XM_021255575.1 XP_021111234.1 14 - 15 1423 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0