This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05263373_g1
See other NSUN5 GE Assays ›
Gene Symbol:
Nsun5
Gene Name:
NOP2/Sun RNA methyltransferase family member 5
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624740: 14413494 - 14418253 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05263373_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nsun5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840076.3 XP_004840133.1 2 - 3 256 67
XM_004840077.3 XP_004840134.1 2 - 3 255 67
XM_021254428.1 XP_021110087.1 2 - 3 277 67
XM_021254429.1 XP_021110088.1 2 - 3 277 67
XM_021254430.1 XP_021110089.1 2 - 3 277 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0