This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05267008_s1
See other ZNF239 GE Assays ›
Gene Symbol:
Znf239
Gene Name:
zinc finger protein 239
Gene Aliases:
GW7_14918
Chromosome Location:
Chr.Un|NW_004624922: 647018 - 649414 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05267008_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Znf239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004921224.3 XP_004921281.2 2 618 95
XM_021251598.1 XP_021107257.1 4 853 95
XM_021251599.1 XP_021107258.1 4 902 95
XM_021251600.1 XP_021107259.1 4 1116 95
XM_021251601.1 XP_021107260.1 4 840 95
XM_021251602.1 XP_021107261.1 3 718 95
XM_021251603.1 XP_021107262.1 3 784 95
XM_021251604.1 XP_021107263.1 3 658 95
XM_021251605.1 XP_021107264.1 3 998 95
XM_021251607.1 XP_021107266.1 3 723 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0