This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00153690_m1
See other AANAT GE Assays ›
Gene Symbol:
AANAT
Gene Name:
aralkylamine N-acetyltransferase
Gene Aliases:
DSPS, SNAT
Chromosome Location:
Chr.17: 76453351 - 76470797 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00153690_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AANAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001088.2 NP_001079.1 1 - 2 180 84
NR_110548.1 1 - 2 180 84
GenBank mRNA AA084486.1 - 1 - 2 179 84
AB593119.1 - 1 - 2 128 84
BC069434.1 - 1 - 2 123 7262229 84
U40347.1 - 1 - 2 159 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008946.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000873.hg.1

Additional Information:

This assay is in the 5'UTR region of the AB593119.1,BC069434.1,NM_001088.2,U40347.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AANAT

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
circadian rhythm
photoperiodism
response to zinc ion
melatonin biosynthetic process
response to insulin
response to cytokine
response to prostaglandin E
indolalkylamine biosynthetic process
response to copper ion
response to corticosterone
response to calcium ion
cellular response to cAMP
aralkylamine N-acetyltransferase activity
arylamine N-acetyltransferase activity
protein binding
14-3-3 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17011347.hg.1 ENST00000392492.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17011347.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011347.hg.1 TC17000873.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17011349.hg.1 ENST00000250615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17011349.hg.1 ENST00000392492.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17011349.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011349.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011349.hg.1 TC17000873.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17011348.hg.1 ENST00000392492.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17011348.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011348.hg.1 TC17000873.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay TC1700008946.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167406.hg.1 TC1700008946.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167401.hg.1 TC1700008946.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167402.hg.1 TC1700008946.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000250615.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000392492.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000585649.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 ENST00000587798.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NM_001088.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NM_001166579.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 NR_110548.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167405.hg.1 TC1700008946.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0