This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00153890_m1
See other ADPRH GE Assays ›
Gene Symbol:
ADPRH
Gene Name:
ADP-ribosylarginine hydrolase
Gene Aliases:
ARH1
Chromosome Location:
Chr.3: 119579433 - 119589954 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00153890_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADPRH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001125.3 NP_001116.1 3 - 4 845 74
NM_001291949.1 NP_001278878.1 2 - 3 759 74
XM_011512438.2 XP_011510740.1 3 - 4 911 74
GenBank mRNA AK291787.1 - 2 - 3 602 74
AK313369.1 - 2 - 3 486 74
BC063883.1 - 3 - 4 602 30374536 74
BC074769.2 - 1 - 2 343 30915190 74
L13291.1 - 1 - 2 409 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008482.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000610.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADPRH

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
intracellular protein transport
regulation of vesicle fusion
protein de-ADP-ribosylation
activation of GTPase activity
magnesium ion binding
ADP-ribosylarginine hydrolase activity
GTPase activator activity
protein binding
Rab GTPase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000357003.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000465513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000471850.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478399.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000478927.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000481816.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 ENST00000485496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001125.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291949.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008482.hg.1 NM_001291950.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 160378 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline