This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00159589_m1
See other NDUFS4 GE Assays ›
Gene Symbol:
NDUFS4
Gene Name:
NADH:ubiquinone oxidoreductase subunit S4
Gene Aliases:
AQDQ, CI-18, CI-18 kDa, CI-AQDQ
Chromosome Location:
Chr.5: 53560610 - 53683341 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00159589_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318051.1 NP_001304980.1 1 - 2 130 66
NM_002495.3 NP_002486.1 1 - 2 130 66
NR_134473.1 1 - 2 130 66
NR_134474.1 1 - 2 130 66
XM_017009491.1 XP_016864980.1 1 - 2 153 66
GenBank mRNA AF020351.1 - 1 - 2 108 66
AK307973.1 - 1 - 2 108 66
BU145342.1 - 1 - 2 104 66
BU954271.1 - 1 - 2 122 66
DA609203.1 - 1 - 2 130 66
DB489661.1 - 1 - 2 129 66
KU178226.1 - 1 - 2 100 66
KU178227.1 - 1 - 2 100 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007378.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000221.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFS4

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
mitochondrial electron transport, NADH to ubiquinone
brain development
cAMP-mediated signaling
mitochondrial respiratory chain complex I assembly
cellular respiration
positive regulation of fibroblast proliferation
response to cAMP
reactive oxygen species metabolic process
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05002747.hg.1 ENST00000296684.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002747.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002747.hg.1 TC05000221.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05002748.hg.1 ENST00000296684.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002748.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002748.hg.1 TC05000221.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05002750.hg.1 ENST00000296684.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002750.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05002750.hg.1 TC05000221.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149893.hg.1 TC0500007378.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149896.hg.1 TC0500007378.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0