This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00173848_m1
See other GPR75 GE Assays ›
Gene Symbol:
GPR75
Gene Name:
G protein-coupled receptor 75
Gene Aliases:
GPRchr2, WI31133
Chromosome Location:
Chr.2: 53852913 - 53860033 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00173848_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006794.3 NP_006785.1 1 - 2 211 75
GenBank mRNA AF072693.1 - 1 - 2 106 73
AK314885.1 - 1 - 2 167 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016668.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02005025.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF072693.1,AK314885.1,NM_006794.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR75

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
regulation of neuron death
G-protein coupled receptor activity
C-C chemokine receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02029486.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029486.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029486.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02029479.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029479.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029479.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029485.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029485.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029485.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029480.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029480.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029480.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02029478.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029478.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029478.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02029484.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02029484.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029484.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02029488.hg.1 NM_006794.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02029488.hg.1 TC02005025.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay TC0200016668.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226317.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226317.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226317.hg.1 TC0200016668.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200226314.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226314.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226314.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226314.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226314.hg.1 TC0200016668.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200226312.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226312.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226312.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226312.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226312.hg.1 TC0200016668.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200226316.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226316.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226316.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226316.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226316.hg.1 TC0200016668.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000406625.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000459916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 ENST00000498475.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay TC0200016667.hg.1 NM_001164165.1 Refseq
Affymetrix Human Clariom D Assay PSR0200226313.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226313.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226313.hg.1 ENST00000394705.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200226313.hg.1 NM_006794.3 Refseq
Affymetrix Human Clariom D Assay PSR0200226313.hg.1 TC0200016668.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline