This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00173855_m1
See other GPR176 GE Assays ›
Gene Symbol:
GPR176
Gene Name:
G protein-coupled receptor 176
Gene Aliases:
HB-954
Chromosome Location:
Chr.15: 39795049 - 39920933 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00173855_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR176
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007223.2 NP_009154.1 1 - 2 1042 65
XM_017021874.1 XP_016877363.1 1 - 2 1077 65
GenBank mRNA D38449.1 - 1 - 2 1042 65
JQ308174.1 - 1 - 2 176 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500010854.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001225.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017021874.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR176

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
chemical synaptic transmission
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15011006.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011006.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011006.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15011006.hg.1 TC15001225.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15010998.hg.1 ENST00000543580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010998.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010998.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010998.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15010998.hg.1 TC15001225.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15011007.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011007.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15011007.hg.1 TC15001225.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 ENST00000543580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15010996.hg.1 TC15001225.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184321.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184337.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184338.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184336.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184323.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay PSR1500184339.hg.1 TC1500010854.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000299092.3 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000543580.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000558041.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000560729.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 ENST00000561100.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271854.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_001271855.1 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq
Affymetrix Human Clariom D Assay TC1500010854.hg.1 NM_007223.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0