This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00184475_m1
See other GSTT1 GE Assays ›
Gene Symbol:
GSTT1
Gene Name:
glutathione S-transferase theta 1
Gene Aliases:
-
Chromosome Location:
Chr.22_KI270879v1: 270308 - 278486 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00184475_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000853.3 NP_000844.2 2 - 3 284 109
GenBank mRNA AF435971.1 - 2 - 3 204 109
BM765626.1 - 2 - 3 257 109
BT019951.1 - 2 - 3 204 109
DA900601.1 - 3 - 4 353 109
X79389.1 - 2 - 3 204 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000583.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GSTT1

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
glutathione peroxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 ENST00000382792.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 ENST00000452369.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010298.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22010286.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010286.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010286.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010286.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22010284.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010284.hg.1 ENST00000382792.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010284.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010284.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010284.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22010291.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010291.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010291.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22010285.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010285.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010285.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010285.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200008204.hg.1 ENST00000437021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008204.hg.1 ENST00000437021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008204.hg.1 ENST00000437021.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008204.hg.1 ENST00000437021.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008204.hg.1 ENST00000437021.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008204.hg.1 ENST00000437021.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0