This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00192725_m1
See other GFPT2 GE Assays ›
Gene Symbol:
GFPT2
Gene Name:
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases:
GFAT, GFAT 2, GFAT2
Chromosome Location:
Chr.5: 180300690 - 180353387 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00192725_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GFPT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005110.3 NP_005101.1 13 - 14 1442 88
XM_006714942.2 XP_006715005.1 13 - 14 1391 88
XM_017010100.1 XP_016865589.1 12 - 13 1327 88
GenBank mRNA AB016789.1 - 13 - 14 1371 88
AK001242.1 - 3 - 4 324 88
AK022507.1 - 13 - 14 1382 88
AK297750.1 - 11 - 12 1078 88
AK314551.1 - 13 - 14 1382 88
BC000012.2 - 13 - 14 1380 3506723 88
BT009818.1 - 12 - 13 1272 88
KU178494.1 - 12 - 13 1272 88
KU178495.1 - 12 - 13 1272 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GFPT2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose 6-phosphate metabolic process
UDP-N-acetylglucosamine metabolic process
UDP-N-acetylglucosamine biosynthetic process
energy reserve metabolic process
glutamine metabolic process
glutamine-fructose-6-phosphate transaminase (isomerizing) activity
protein binding
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000253778.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000503546.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000510122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518158.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518185.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000518906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 ENST00000520165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500013089.hg.1 NM_005110.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline