This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00193376_m1
See other GJB5 GE Assays ›
Gene Symbol:
GJB5
Gene Name:
gap junction protein beta 5
Gene Aliases:
CX31.1
Chromosome Location:
Chr.1: 34754777 - 34758513 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00193376_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005268.3 NP_005259.1 1 - 2 147 135
GenBank mRNA AF052693.1 - 1 - 2 141 135
AK129509.1 - 1 - 2 147 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007745.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000448.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF052693.1,NM_005268.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJB5

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

cell communication
epidermis development
trophoblast giant cell differentiation
spongiotrophoblast differentiation
labyrinthine layer morphogenesis

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01006935.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006935.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006935.hg.1 TC01000448.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006933.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01006933.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01006933.hg.1 TC01000448.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154541.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154541.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154541.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154541.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154541.hg.1 TC0100007745.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100154542.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154542.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154542.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154542.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154542.hg.1 TC0100007745.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100154544.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154544.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154544.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100154544.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154544.hg.1 TC0100007745.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline