This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00199111_m1
See other MLLT11 GE Assays ›
Gene Symbol:
MLLT11
Gene Name:
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
Gene Aliases:
AF1Q
Chromosome Location:
Chr.1: 151059675 - 151068497 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00199111_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLLT11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006818.3 NP_006809.1 1 - 2 883 142
GenBank mRNA AK056089.1 - 1 - 2 878 142
AK057755.1 - 1 - 2 153 142
BC007079.1 - 1 - 2 147 142
BC008445.1 - 1 - 2 147 4103485 142
BC009624.1 - 1 - 2 163 3878959 142
BC019253.1 - 1 - 2 123 3502660 142
BC021703.1 - 1 - 2 221 4108071 142
BC022448.1 - 1 - 2 1155 4815214 142
BC032996.1 - 1 - 2 180 142
U16954.1 - 1 - 2 353 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009938.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001201.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK056089.1,BC008445.1,BC009624.1,BC019253.1,BC021703.1,BC022448.1,NM_006818.3,U16954.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MLLT11

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of mitochondrial depolarization
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway
intrinsic apoptotic signaling pathway
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009938.hg.1 NM_006818.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000357235.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000470278.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000483763.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000491825.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000492796.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 ENST00000540998.5 Ensembl
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_001038707.1 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay TC0100018486.hg.1 NM_020239.3 Refseq
Affymetrix Human Clariom D Assay PSR0100171890.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100171890.hg.1 ENST00000368921.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100171890.hg.1 TC0100009938.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 151049 - -
ABI Human Genome Survey Microarray Version 2.0 225417 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0