| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300182928.hg.1 |
TC0300011124.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000348721.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000443053.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
ENST00000491847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_013324.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011124.hg.1 |
NM_145071.2 |
Refseq |