This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00204120_m1
See other TMPRSS11E GE Assays ›
Gene Symbol:
TMPRSS11E
Gene Name:
transmembrane protease, serine 11E
Gene Aliases:
DESC1, TMPRSS11E2
Chromosome Location:
Chr.4: 68447449 - 68497604 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00204120_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS11E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014058.3 NP_054777.2 1 - 2 73 80
GenBank mRNA AF064819.1 - 1 - 2 61 80
AK292787.1 - 1 - 2 59 80
AK313859.1 - 1 - 2 73 80
AY359017.1 - 1 - 2 40 80
DB013544.1 - 1 - 2 73 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400007734.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000369.hg.1, TC4_ctg9_hap1000001.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF064819.1,AK292787.1,AK313859.1,AY359017.1,NM_014058.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMPRSS11E

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cognition
serine-type endopeptidase activity
serine-type peptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000002.hg.1 TC4_ctg9_hap1000001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000002.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000001.hg.1 TC4_ctg9_hap1000001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000001.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04004998.hg.1 TC04000369.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04004998.hg.1 ENST00000305363.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04004998.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04004999.hg.1 TC04000369.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04004999.hg.1 ENST00000305363.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04004999.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000003.hg.1 TC4_ctg9_hap1000001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR4_ctg9_hap1000003.hg.1 NM_014058.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04004997.hg.1 TC04000369.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04004997.hg.1 ENST00000305363.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04004997.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152692.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400152692.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152692.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400152692.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152692.hg.1 TC0400007734.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay TC0400007734.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 ENST00000305363.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 ENST00000510647.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 NM_014058.3 Refseq
Affymetrix Human Clariom D Assay PSR0400152695.hg.1 TC0400007734.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0