This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00205363_m1
See other IL37 GE Assays ›
Gene Symbol:
IL37
Gene Name:
interleukin 37
Gene Aliases:
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
Chromosome Location:
Chr.2: 112908889 - 112918882 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00205363_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173205.1 NP_775297.1 1 - 2 69 84
GenBank mRNA AF201832.1 - 1 - 2 69 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008942.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000714.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL37

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
immune response
cytokine-mediated signaling pathway
cytokine activity
interleukin-1 receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02011087.hg.1 TC02000714.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02011087.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02011087.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay PSR0200162446.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200162446.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay PSR0200162446.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200162446.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay PSR0200162446.hg.1 TC0200008942.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000263326.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000311328.2 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000349806.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000352179.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 ENST00000353225.7 Ensembl
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_014439.3 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173202.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173203.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173204.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173205.1 Refseq
Affymetrix Human Clariom D Assay TC0200008942.hg.1 NM_173205.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0