This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00210582_m1
See other GTF2A1 GE Assays ›
Gene Symbol:
GTF2A1
Gene Name:
general transcription factor IIA subunit 1
Gene Aliases:
TF2A1, TFIIA, TFIIA-42, TFIIAL
Chromosome Location:
Chr.14: 81175452 - 81221231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00210582_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278940.1 NP_001265869.1 1 - 2 466 103
NM_015859.3 NP_056943.1 1 - 2 466 103
GenBank mRNA BC107155.2 - 1 - 2 201 40032646 103
BC107156.1 - 1 - 2 203 40032650 103
CB240876.1 - 2 - 3 103
D14887.1 - 1 - 2 436 103
X75383.1 - 1 - 2 173 103
X77225.1 - 1 - 2 32 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009834.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001368.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001278940.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Transcription of mRNA 1700 Inventoried MicroArray Quality Control (MAQC) Human_Transcription_of_mRNA Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2A1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
TBP-class protein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000298173.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000434192.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000553612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 ENST00000556268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_001278940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_015859.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009834.hg.1 NM_201595.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 117851 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0